accessibility menu, dialog, popup

UserName

CAS RN 1140909-48-3

IUPAC Name: 2-hydroxybutanedioic acid; N'1-{4-[(6,7-dimethoxyquinolin-4-yl)oxy]phenyl}-N1-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide
Synonyms: cabozantinib; malic acid
Molecular Weight (g/mol): 635.6
Molecular Formula: C32H30FN3O10
InChi Key: HFCFMRYTXDINDK-UHFFFAOYNA-N
SMILES: OC(CC(O)=O)C(O)=O.COC1=C(OC)C=C2C(OC3=CC=C(NC(=O)C4(CC4)C(=O)NC4=CC=C(F)C=C4)C=C3)=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results