missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CA 074

Catalog No. 48631
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48631 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48631 Supplier R&D Systems™ Supplier No. 4863/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Selective cathepsin B inhibitor

Inhibitor of cathepsin B (Ki = 2-5 nM). Displays selectivity over cathepsins H and L (Ki = 40-200 μM). Shown to reduce bone metastasis in a 4T1.2 breast cancer model.

Chemical Identifiers

CAS 134448-10-5
Molecular Formula C18H29N3O6

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 383.44
Purity >98%
Molecular Weight (g/mol) 383.45
Chemical Name or Material CA 074
Recommended Storage Store at -20°C