missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals C6 Ceramide

CAS: 124753-97-5 | C24H47NO3, C24H47NO3 | 397.64 g/mol

Supplier:  Thermo Scientific Chemicals J67034MA

Quantity 10 mg

 View more versions of this product

Catalog No. AAJ67034MA


Only null left

Description

Description

C-6 ceramide is a cell-permeable analog of naturally occurring ceramides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124753-97-5
C24H47NO3
N-Hexanoyl-D-erythro-sphingosine;

Specifications

10 mg
-30°C to -10°C
C24H47NO3
N-Hexanoyl-D-erythro-sphingosine;
Crystalline solid
CCCCCCCCCCCCC\C=C\C(O)C(CO)NC(=O)CCCCC
397.64
White
124753-97-5
397.64
C6 Ceramide
NPRJSFWNFTXXQC-VHEBQXMUNA-N
N-[(4E)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions