accessibility menu, dialog, popup

UserName

H3COHHOHNOCH3(E)

CAS RN 124753-97-5

IUPAC Name: N-[(4E)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
Synonyms: caproyl sphingosine
Molecular Weight (g/mol): 397.64
Molecular Formula: C24H47NO3
InChi Key: NPRJSFWNFTXXQC-VHEBQXMUNA-N
SMILES: CCCCCCCCCCCCC\C=C\C(O)C(CO)NC(=O)CCCCC
Product Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results