missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals C6 Ceramide

Catalog No. p-7070111
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67034MCR 1 mg
AAJ67034LB0 5 mg
AAJ67034MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67034MCR Supplier Thermo Scientific Chemicals Supplier No. J67034MCR
Only null left

CAS: 124753-97-5 | C24H47NO3, C24H47NO3 | 397.64 g/mol

C-6 ceramide is a cell-permeable analog of naturally occurring ceramides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124753-97-5
Molecular Formula C24H47NO3
Synonym N-Hexanoyl-D-erythro-sphingosine;

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
Formula Weight 397.64
Product Type C6 Ceramide
Form Crystalline solid
InChI Key NPRJSFWNFTXXQC-VHEBQXMUNA-N
SMILES CCCCCCCCCCCCC\C=C\C(O)C(CO)NC(=O)CCCCC
IUPAC Name N-[(4E)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
Molecular Weight (g/mol) 397.64
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only