missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3orf38 Recombinant Rabbit Monoclonal Antibody (23GB1150)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Specifications
| Antigen | C3orf38 |
|---|---|
| Clone | 23GB1150 |
| Concentration | 0.6 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
C3orf38 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Specifications
| C3orf38 | |
| 0.6 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5JPI3 | |
| 100503356, 285237 | |
| A synthesized peptide derived from human C3orf38 (51-200AA). | |
| Antibody |
| 23GB1150 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| C3orf38 | |
| C3orf38; chromosome 3 open reading frame 38; uncharacterized protein C3orf38 | |
| C3orf38, Gm19653 | |
| Primary | |
| Affinity chromatography |