missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C3orf38 Recombinant Rabbit Monoclonal Antibody (23GB1150)

Catalog No. MA553345
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553345 20 μL
MA553344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553345 Supplier Invitrogen™ Supplier No. MA553345
Only null left

Rabbit Recombinant Monoclonal Antibody

C3orf38 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Specifications

Antigen C3orf38
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1150
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C3orf38
Gene Accession No. Q5JPI3
Gene Alias C3orf38; chromosome 3 open reading frame 38; uncharacterized protein C3orf38
Gene Symbols C3orf38, Gm19653
Host Species Rabbit
Immunogen A synthesized peptide derived from human C3orf38 (51-200AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100503356, 285237
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less