missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ C16

Catalog No. 538250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
538250 50 mg
538210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 538250 Supplier Tocris Bioscience™ Supplier No. 5382/50
Only null left

PKR inhibitor

C16 is a PKR inhibitor (IC50 = 210 nM). Rescues PKR-dependent translation block in vitro. Decreases Aβ42-induced inflammatory cytokine release and apoptosis in neuronal cultures. Prevents neuroinflammation and neuronal loss in an acute excitotoxic rat model. Brain penetrant.

Specifications

Chemical Name or Material C16
CAS 608512-97-6
Synonym 6,8-Dihydro-8-(1H-imidazol-5-ylmethylene)-7H-pyrrolo[2,3-g]benzothiazol-7-one
Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 20mM in ethanol
Molecular Formula C13H8N4OS
Formula Weight 268.29
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use