missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ C-Laurdan
Description
C-Laurdan is a fluorescent probe for imaging lipid rafts. Used to detect membrane polarity via one and two-photon fluorecence imaging. Two photon excitation spectra = 780 nm; one photon absorption/emission spectra = 348/423 nm; Quantum yield = 0.43.
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-[6-(1-oxododecyl)-2-naphthalenyl]glycine |
| CAS | 959839-06-6 |
| Quantity | 2 mg |
| Product Line | Tocris |
| Target | Fluorescent Lipids Probes and Cell Membrane Stains |
| Molecular Formula | C25H35NO3 |
| Purity | 0.95 |