missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylsuccinic Anhydride 98.0+%, TCI America™
$277.23 - $831.67
Chemical Identifiers
| CAS | 2035-76-9 |
|---|---|
| Molecular Formula | C8H12O3 |
| Molecular Weight (g/mol) | 156.181 |
| InChI Key | NSJAWXMCLJVBPM-UHFFFAOYSA-N |
| PubChem CID | 235910 |
| IUPAC Name | 3-butyloxolane-2,5-dione |
| SMILES | CCCCC1CC(=O)OC1=O |
Chemical Identifiers
| 2035-76-9 | |
| 156.181 | |
| 235910 | |
| CCCCC1CC(=O)OC1=O |
| C8H12O3 | |
| NSJAWXMCLJVBPM-UHFFFAOYSA-N | |
| 3-butyloxolane-2,5-dione |
Specifications
| 46°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| NSJAWXMCLJVBPM-UHFFFAOYSA-N | |
| 3-butyloxolane-2,5-dione | |
| 235910 | |
| ≥98.0% (GC) |
| 2035-76-9 | |
| 5 g | |
| C8H12O3 | |
| CCCCC1CC(=O)OC1=O | |
| 156.181 | |
| 156.18 | |
| Butylsuccinic Anhydride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator