accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 2035-76-9

IUPAC Name: 3-butyloxolane-2,5-dione
Synonyms: 3-butyloxolane-2,5-dione
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: NSJAWXMCLJVBPM-UHFFFAOYNA-N
SMILES: CCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results