missing translation for 'onlineSavingsMsg'
Learn More

Butylsuccinic Anhydride 98.0+%, TCI America™

Catalog No. b27425g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B27425G 5 g
B274225G 25 g
2 options

Catalog No. B27425G

Supplier: TCI America B27425G

Only null left

Chemical Identifiers

CAS 2035-76-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
InChI Key NSJAWXMCLJVBPM-UHFFFAOYSA-N
PubChem CID 235910
IUPAC Name 3-butyloxolane-2,5-dione
SMILES CCCCC1CC(=O)OC1=O

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material Butylsuccinic Anhydride
TSCA No
Recommended Storage Refrigerator