missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Butyl Sepharose™ High Performance Chromatography Media

Designed for intermediate and polishing step purification steps

$1,992.25

Specifications

For Use With (Application) For Intermediate and Polishing Step PurifiCatIon Steps
Stability Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS
Storage Requirements 4°C to 30°C
Binding Capacity 38 mg b-lactoglobulin/mL mediμm
Product Type Butyl Sepharose High Performance Media
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45002955
View Documents Promotion Details
Cytiva
17543202
200 mL
Each for $1,992.25
Only null left
 
Description

Description

  • Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked 34μm agarose beads with aliphatic butyl functional groups
  • High-resolution, capacity separations for recovery
  • High ligand density for dynamic binding capacity even at low salt concentration
  • Display minimum of ionic interactions
  • Chemical stability for effective CIP and sanitization
  • Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes

Specifications

Specifications

For Intermediate and Polishing Step PurifiCatIon Steps
4°C to 30°C
Butyl Sepharose High Performance Media
24 to 44 μm
Cross-Linked Agarose
20% Ethanol
Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS
38 mg b-lactoglobulin/mL mediμm
100 to 200 cm/Hoμr
300 kPa
3 to 13
Butyl
Videos
SDS
Documents

Documents