missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Butyl Sepharose™ High Performance Chromatography Media

Catalog No. 45002955
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002955 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002955 Supplier Cytiva Supplier No. 17543202

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for intermediate and polishing step purification steps

  • Hydrophobic interaction chromatography (HIC) media composed of highly cross-linked 34μm agarose beads with aliphatic butyl functional groups
  • High-resolution, capacity separations for recovery
  • High ligand density for dynamic binding capacity even at low salt concentration
  • Display minimum of ionic interactions
  • Chemical stability for effective CIP and sanitization
  • Available in prepacked columns such as HiPrep™, HiScreen™, and HiTrap™ plus laboratory and BioProcess pack sizes

Specifications

Quantity 200 mL
For Use With (Application) For Intermediate and Polishing Step PurifiCatIon Steps
Stability Stable in commonly used aqueous Buffers: 1 M Sodium Hydroxide, 1 M Acetic Acid, 8 M Urea, 6 M Guanidine Hydrochloride, 30% Acetonitrile, 70% Ethanol, 3 M Ammonium sulfate, 1 mM HCl, 30% Isopropanol, 2% SDS
Storage Requirements 4°C to 30°C
Binding Capacity 38 mg b-lactoglobulin/mL mediμm
Product Type Butyl Sepharose High Performance Media
Flow Rate 100 to 200 cm/Hoμr
Particle Size 24 to 44 μm
Max. Pressure 300 kPa
Matrix Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 20% Ethanol
Ligand Type Butyl
Show More Show Less