missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Salicylate 99.0+%, TCI America™
$50.33 - $251.65
Chemical Identifiers
| CAS | 2052-14-4 |
|---|---|
| Molecular Formula | C11H14O3 |
| Molecular Weight (g/mol) | 194.23 |
| MDL Number | MFCD00020038 |
| InChI Key | YFDUWSBGVPBWKF-UHFFFAOYSA-N |
| Synonym | Salicylic Acid Butyl Ester, 2-Hydroxybenzoic Acid Butyl Ester, Butyl 2-Hydroxybenzoate |
| PubChem CID | 16330 |
| IUPAC Name | butyl 2-hydroxybenzoate |
| SMILES | CCCCOC(=O)C1=CC=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S001025ML
|
TCI America
S001025ML |
25 mL |
Each for $50.33
|
|
|||||
|
S0010500ML
|
TCI America
S0010500ML |
500 mL |
Each for $251.65
|
|
|||||
Chemical Identifiers
| 2052-14-4 | |
| 194.23 | |
| YFDUWSBGVPBWKF-UHFFFAOYSA-N | |
| 16330 | |
| CCCCOC(=O)C1=CC=CC=C1O |
| C11H14O3 | |
| MFCD00020038 | |
| Salicylic Acid Butyl Ester, 2-Hydroxybenzoic Acid Butyl Ester, Butyl 2-Hydroxybenzoate | |
| butyl 2-hydroxybenzoate |
Specifications
| 2052-14-4 | |
| 260°C | |
| MFCD00020038 | |
| Salicylic Acid Butyl Ester, 2-Hydroxybenzoic Acid Butyl Ester, Butyl 2-Hydroxybenzoate | |
| CCCCOC(=O)C1=CC=CC=C1O | |
| 194.23 | |
| 194.23 | |
| Liquid |
| Colorless | |
| C11H14O3 | |
| 25 mL | |
| YFDUWSBGVPBWKF-UHFFFAOYSA-N | |
| butyl 2-hydroxybenzoate | |
| 16330 | |
| ≥99.0% (GC) | |
| Butyl Salicylate |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : VO2000000
TSCA : Yes