accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 2052-14-4

IUPAC Name: butyl 2-hydroxybenzoate
Synonyms: butyl salicylate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: YFDUWSBGVPBWKF-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=CC=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results