missing translation for 'onlineSavingsMsg'
Learn More

Butyl Salicylate 99.0+%, TCI America™

Catalog No. S001025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001025ML 25 mL
S0010500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001025ML Supplier TCI America Supplier No. S001025ML

Chemical Identifiers

CAS 2052-14-4
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00020038
InChI Key YFDUWSBGVPBWKF-UHFFFAOYSA-N
Synonym Salicylic Acid Butyl Ester, 2-Hydroxybenzoic Acid Butyl Ester, Butyl 2-Hydroxybenzoate
PubChem CID 16330
IUPAC Name butyl 2-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=CC=C1O

Specifications

Color Colorless
Boiling Point 260°C
Quantity 25 mL
Formula Weight 194.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO2000000
TSCA Yes