missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Ricinoleate 74.0+%, TCI America™
Supplier: TCI America R0045500ML
Specifications
| Butyl Ricinoleate | |
| Yellow | |
| C22H42O3 | |
| 500 mL | |
| HGWAKQDTQVDVRP-QINSGFPZNA-N | |
| butyl (9Z)-12-hydroxyoctadec-9-enoate | |
| 40468183 | |
| ≥74.0% (GC) |
| 151-13-3 | |
| 275°C | |
| MFCD00136031 | |
| Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester | |
| CCCCCCC(O)C\C=C/CCCCCCCC(=O)OCCCC | |
| 354.58 | |
| 354.58 | |
| Liquid |
Chemical Identifiers
| 151-13-3 | |
| 354.58 | |
| HGWAKQDTQVDVRP-QINSGFPZNA-N | |
| 40468183 | |
| CCCCCCC(O)C\C=C/CCCCCCCC(=O)OCCCC |
| C22H42O3 | |
| MFCD00136031 | |
| Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester | |
| butyl (9Z)-12-hydroxyoctadec-9-enoate |
Safety and Handling
TSCA : Yes