missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Ricinoleate 74.0+%, TCI America™
$29.83 - $87.60
Chemical Identifiers
| CAS | 151-13-3 |
|---|---|
| Molecular Formula | C22H42O3 |
| Molecular Weight (g/mol) | 354.58 |
| MDL Number | MFCD00136031 |
| InChI Key | HGWAKQDTQVDVRP-QINSGFPZNA-N |
| Synonym | Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester |
| PubChem CID | 40468183 |
| IUPAC Name | butyl (9Z)-12-hydroxyoctadec-9-enoate |
| SMILES | CCCCCCC(O)C\C=C/CCCCCCCC(=O)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R004525ML
|
TCI America
R004525ML |
25 mL |
Each for $29.83
|
|
|||||
|
R0045500ML
|
TCI America
R0045500ML |
500 mL |
Each for $87.60
|
|
|||||
Chemical Identifiers
| 151-13-3 | |
| 354.58 | |
| HGWAKQDTQVDVRP-QINSGFPZNA-N | |
| 40468183 | |
| CCCCCCC(O)C\C=C/CCCCCCCC(=O)OCCCC |
| C22H42O3 | |
| MFCD00136031 | |
| Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester | |
| butyl (9Z)-12-hydroxyoctadec-9-enoate |
Specifications
| 151-13-3 | |
| 275°C | |
| MFCD00136031 | |
| Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester | |
| CCCCCCC(O)C\C=C/CCCCCCCC(=O)OCCCC | |
| 354.58 | |
| 354.58 | |
| Liquid |
| Yellow | |
| C22H42O3 | |
| 25 mL | |
| HGWAKQDTQVDVRP-QINSGFPZNA-N | |
| butyl (9Z)-12-hydroxyoctadec-9-enoate | |
| 40468183 | |
| ≥74.0% (GC) | |
| Butyl Ricinoleate |
Safety and Handling
TSCA : Yes