missing translation for 'onlineSavingsMsg'
Learn More

Butyl Propionate 99.0+%, TCI America™

Supplier:  TCI America P050225ML

 View more versions of this product

Catalog No. P050225ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Butyl Propionate
Colorless
C7H14O2
25 mL
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
CCCCOC(=O)CC
130.187
130.19
Liquid
590-01-2
146°C
MFCD00009448
1914
BTMVHUNTONAYDX-UHFFFAOYSA-N
butyl propanoate
11529
≥99.0% (GC)

Chemical Identifiers

590-01-2
130.187
BTMVHUNTONAYDX-UHFFFAOYSA-N
11529
CCCCOC(=O)CC
C7H14O2
MFCD00009448
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
butyl propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0774

RTECSNumber : UE8245000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions