missing translation for 'onlineSavingsMsg'
Learn More

Butyl Propionate 99.0+%, TCI America™

Catalog No. p-7134642
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050225ML 25 mL
P0502500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050225ML Supplier TCI America Supplier No. P050225ML
Only null left

Chemical Identifiers

CAS 590-01-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009448
InChI Key BTMVHUNTONAYDX-UHFFFAOYSA-N
Synonym butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
PubChem CID 11529
IUPAC Name butyl propanoate
SMILES CCCCOC(=O)CC

Specifications

Color Colorless
Boiling Point 146°C
Quantity 25 mL
UN Number 1914
Formula Weight 130.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Propionate
EINECSNumber (2)-0774
RTECSNumber UE8245000
TSCA Yes