accessibility menu, dialog, popup

UserName

CAS RN 590-01-2

IUPAC Name: butyl propanoate
Synonyms: butyl propionate N-butyl propionate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: BTMVHUNTONAYDX-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)