missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methyl Sulfide 98.0+%, TCI America™

Catalog No. B07295ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B07295ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B07295ML Supplier TCI America Supplier No. B07295ML

Chemical Identifiers

CAS 628-29-5
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00015256
InChI Key WCXXISMIJBRDQK-UHFFFAOYSA-N
Synonym butyl methyl sulfide, butane, 1-methylthio, n-butyl methyl sulfide, butyl methyl sulphide, sulfide, butyl methyl, 1-methylthio butane, unii-2n5imb9xap, methyl butyl sulfide, 2-thiahexane, 2n5imb9xap
PubChem CID 12339
IUPAC Name 1-(methylsulfanyl)butane
SMILES CCCCSC

Specifications

Color Colorless
Boiling Point 123°C
Quantity 5 mL
UN Number 1993
Formula Weight 104.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Methyl Sulfide