CAS RN 628-29-5
IUPAC Name:
1-(methylsulfanyl)butane
Synonyms:
2-thiahexane
Molecular Weight (g/mol):
104.21
Molecular Formula:
C5H12S
InChi Key:
WCXXISMIJBRDQK-UHFFFAOYSA-N
SMILES:
CCCCSC
1
–
1
of
1
results
1
Butyl Methyl Sulfide 98.0+%, TCI America™
CAS: 628-29-5 Molecular Formula: C5H12S Molecular Weight (g/mol): 104.21 MDL Number: MFCD00015256 InChI Key: WCXXISMIJBRDQK-UHFFFAOYSA-N Synonym: butyl methyl sulfide,butane, 1-methylthio,n-butyl methyl sulfide,butyl methyl sulphide,sulfide, butyl methyl,1-methylthio butane,unii-2n5imb9xap,methyl butyl sulfide,2-thiahexane,2n5imb9xap PubChem CID: 12339 IUPAC Name: 1-(methylsulfanyl)butane SMILES: CCCCSC