missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
Bulk pack of wide-mouth HDPE reagent bottles with polypropylene caps are chemically resistant, leak-tested, and easy to fill and handle, making them ideal for high-volume lab, industrial, or sample storage applications.
$968.75 - $1,614.57
Specifications
| Color | Translucent |
|---|---|
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
Description
- Bulk pack: Includes bulk pack of wide-mouth HDPE reagent bottles with polypropylene caps, designed for safe liquid storage and lab applications.
- Designed for versatility: Wide-mouth design makes filling and pouring liquids easy, ensuring convenience for lab, industrial, or sample storage applications.
- Chemical resistance: HDPE construction offers excellent resistance to a variety of chemicals, making these bottles suitable for diverse laboratory uses. Designed for secure liquid storage with a -100 to 120°C temperature range.
- Leak-proof tested: Ensures safe transport and storage with rigorously tested caps and bottle threads to prevent leaks, even under pressure.
- Lightweight and translucent: Lightweight HDPE material allows for easy handling, while the translucent body offers visibility of contents.
Specifications
| Translucent | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| HDPE | |
| Yes | |
| Wide |