missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
Description
- Bulk pack: Includes bulk pack of wide-mouth HDPE reagent bottles with polypropylene caps, designed for safe liquid storage and lab applications.
- Designed for versatility: Wide-mouth design makes filling and pouring liquids easy, ensuring convenience for lab, industrial, or sample storage applications.
- Chemical resistance: HDPE construction offers excellent resistance to a variety of chemicals, making these bottles suitable for diverse laboratory uses. Designed for secure liquid storage with a -100 to 120°C temperature range.
- Leak-proof tested: Ensures safe transport and storage with rigorously tested caps and bottle threads to prevent leaks, even under pressure.
- Lightweight and translucent: Lightweight HDPE material allows for easy handling, while the translucent body offers visibility of contents.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4.22 oz. |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 28.8 mm |
| Material | HDPE |
| No. per Pack | 500 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |