missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Bulk Pack of Narrow Mouth HDPE Amber Reagent Bottles with Polypropylene Caps
Bulk pack of narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemically resistant, ideal for storing light-sensitive liquids in lab or industrial environments.
Specifications
| Color | Amber |
|---|---|
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
Description
- Bulk pack: Narrow-mouth amber HDPE reagent bottles with polypropylene caps, designed for secure storage and transport of sensitive liquids and samples.
- Light shielding design: Amber color protects light-sensitive samples and reagents from UV exposure, making these bottles ideal for lab and industrial applications.
- Excellent chemical resistance: HDPE material withstands exposure to various chemicals, ensuring reliable use in laboratory and industrial settings.
- Leak-proof tested: Rigorously tested caps and bottle threads ensure no leaks, providing safe and secure storage for liquids and reagents.
- Lightweight and durable: Made from lightweight, durable HDPE, these translucent amber bottles allow for visibility of contents while providing protection and ease of use. Non-sterile and not autoclavable. Protects light-sensitive substances with a temperature range of -100 to +120°C.
Specifications
| Amber | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| HDPE | |
| Yes | |
| Narrow | |
| Not Autoclavable |