missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Bulk Pack of Narrow Mouth HDPE Amber Reagent Bottles with Polypropylene Caps

Catalog No. S000109
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000109 250 mL
S000110 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000109 Supplier Eisco Labs Supplier No. CH5437PK250

Bulk pack of narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemically resistant, ideal for storing light-sensitive liquids in lab or industrial environments.

  • Bulk pack: Narrow-mouth amber HDPE reagent bottles with polypropylene caps, designed for secure storage and transport of sensitive liquids and samples.
  • Light shielding design: Amber color protects light-sensitive samples and reagents from UV exposure, making these bottles ideal for lab and industrial applications.
  • Excellent chemical resistance: HDPE material withstands exposure to various chemicals, ensuring reliable use in laboratory and industrial settings.
  • Leak-proof tested: Rigorously tested caps and bottle threads ensure no leaks, providing safe and secure storage for liquids and reagents.
  • Lightweight and durable: Made from lightweight, durable HDPE, these translucent amber bottles allow for visibility of contents while providing protection and ease of use. Non-sterile and not autoclavable. Protects light-sensitive substances with a temperature range of -100 to +120°C.

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Amber
Diameter (Metric) Inner Neck 17.6 mm
Material HDPE
No. per Pack 250 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 133 mm
Product Type Reagent Bottle
Diameter (Metric) 61.7 mm
Autoclavable Not Autoclavable
Show More Show Less