missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 9.18 at 25°C, Certified, LabChem™
Supplier: LabChem LC124505
Specifications
| Buffer Solution pH 9.18 at 25°C | |
| Colorless | |
| Boron | |
| Liquid | |
| Passes Test | |
| Cubitainer™ | |
| Soluble in water | |
| Certified |
| 1303-96-4,7732-18-5 | |
| 99.62,0.38 | |
| 9.18 ±0.01 | |
| 20 L | |
| Na2B4O7·10H2O | |
| Traceable to NIST | |
| 381.37 |