missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 9.18 at 25°C, Certified, LabChem™

Catalog No. p-5379488
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC124501 500 mL Poly Bottle
LC124502 1 L Poly Bottle
LC124504 4 L Poly Bottle
LC124506 10 L Cubitainer™
LC124505 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC124501 Supplier LabChem Supplier No. LC124501
Only null left

Specifications

Chemical Name or Material Buffer Solution pH 9.18 at 25°C
CAS 1303-96-4, 7732-18-5
Color Colorless
CAS Max % 99.62, 0.38
Decomposition Information Boron
pH 9.18 ±0.01
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula Na2B4O7·10H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 381.37
Grade Certified
Show More Show Less