missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 3.5 R, Ricca Chemical
$253.50 - $430.50
Specifications
| Chemical Name or Material | Buffer Solution |
|---|---|
| Concentration | 1X |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 3.5 |
Specifications
| Buffer Solution | |
| Calibration | |
| 3.5 | |
| pH Buffer |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| Natural Poly Bottle |