missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution for Fluoride, Certified, LabChem™
Supplier: LabChem LC125804
Specifications
| Colorless | |
| Liquid | |
| Poly Bottle |
| 5.0 to 5.5 | |
| 4 L | |
| Soluble in Water |