missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution for Fluoride, Certified, LabChem™

Catalog No. p-5388052
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC125802 1 L
LC125804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC125802 Supplier LabChem Supplier No. LC125802
Only null left

Specifications

Chemical Name or Material Buffer Solution for Fluoride
CAS 6132-04-3, 7647-14-5, 64-19-7, 1310-73-2, 6132-04-3
Color Colorless
CAS Max % 84.07, 5.8, 5.7, 4.4, 0.03
Decomposition Information Carbon monoxide; Carbon dioxide
pH 5.0 to 5.5
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Assay Percent Notes in-process pH
Molecular Formula C6H9Na3O9
Linear Formula NaCl
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Molecular Weight (g/mol) 294.10
Formula Weight 58.44
Grade Certified
Show More Show Less