missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20°C), Honeywell Fluka™
Borax / hydrochloric acid, traceable to SRM from NIST
$156.13 - $1067.39
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1336-21-6 | |
| Sodium tetraborate buffer solution |
Specifications
| 1336-21-6 | |
| 500 mL | |
| MFCD00066650 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| azanium;hydroxide | |
| 14923 | |
| Buffer solution |
| 9.0 | |
| H5NO | |
| NONH for all modes of transport | |
| N.O | |
| 35.05 | |
| CHEBI:18219 |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
Recommended Storage : Room Temp