missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 9.0 (20°C), Buffer solution pH 9.0 (20°C), Honeywell Fluka™

Borax / hydrochloric acid, traceable to SRM from NIST

Supplier:  Honeywell Inc 336481L

 View more versions of this product

Catalog No. 6002456


Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Buffer solution
9.0
H5NO
NONH for all modes of transport
N.O
35.05
CHEBI:18219
1336-21-6
1 L
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923

Chemical Identifiers

1336-21-6
35.05
14923
N.O
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
CHEBI:18219
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions