missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Honeywell Fluka™

With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

$155.66 - $1063.05

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002447
View Documents
Honeywell Chemicals
33643500ML
500 mL
Each for $155.66
Only null left
 
6002446
View Documents
Honeywell Chemicals
336431L
1 L
Each for $204.29
Only null left
 
6002448
View Documents
Honeywell Chemicals
336435LVP
5 L
Each for $486.61
Only null left
 
60046287
View Documents
Honeywell Chemicals
336436X1L
6 x 1 L
Case of 6 Each for $1,063.05
Only null left
 
6002445
View Documents
Honeywell Chemicals
3364310LVP
10 L
Each for $655.34
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
CHEBI:18219
N.O
H5NO
MFCD00066650
14923
azanium;hydroxide

Specifications

1336-21-6
500 mL
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923
Buffer solution
4.0
H5NO
NONH for all modes of transport
N.O
35.05
CHEBI:18219
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P337 + P313

SDS
Documents

Documents