missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Honeywell Fluka™

With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

Supplier:  Honeywell Chemicals 3364310LVP

 View more versions of this product

Catalog No. 6002445


Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Buffer solution
4.0
H5NO
NONH for all modes of transport
N.O
35.05
CHEBI:18219
1336-21-6
10 L
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
CHEBI:18219
N.O
H5NO
MFCD00066650
14923
azanium;hydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338-P337 + P313

SDS
Documents

Documents

Promotions

Promotions