missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, 12.0, di-sodium hydrogen phosphate / sodium hydroxide solution, Honeywell Fluka™
di-Sodium hydrogen phosphate / sodium hydroxide solution
$110.97 - $242.37
Chemical Identifiers
| CAS | 1336-21-6 |
|---|---|
| Molecular Formula | H5NO |
| Molecular Weight (g/mol) | 35.05 |
| MDL Number | MFCD00066650 |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| PubChem CID | 14923 |
| ChEBI | CHEBI:18219 |
| IUPAC Name | azanium;hydroxide |
| SMILES | N.O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| CHEBI:18219 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| 14923 | |
| azanium;hydroxide |
Specifications
| 1336-21-6 | |
| 12.0 | |
| H5NO | |
| NONH for all modes of transport | |
| N.O | |
| 35.05 | |
| CHEBI:18219 |
| 1.00g/mL at 20°C | |
| 100 mL | |
| MFCD00066650 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| azanium;hydroxide | |
| 14923 | |
| Buffer solution |