missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, 12.0, di-sodium hydrogen phosphate / sodium hydroxide solution, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Density | 1.00g/mL at 20°C |
| pH | 12.0 |
| Quantity | 1 L |
| UN Number | NONH for all modes of transport |
| Chemical Name or Material | Buffer solution |