missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 3.00 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 14951

 View more versions of this product

Catalog No. 1495-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer, Reference Standard
877-24-7 , 7732-18-5
Calibration
3
4 L
Natural Poly Bottle
pH 3.00 ± 0.01 at 25°C
1X
pH Meter and Electrode
Liquid
pH Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions