missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical
$42.00 - $431.20
Specifications
| CAS Min % | 97.0,1.02,0.11 |
|---|---|
| CAS Max % | 100.0,1.06,0.11 |
| Concentration | 1X |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R161016
|
Ricca Chemical Company
161016 |
500 mL | Clear PET-G Bottle |
Each for $42.00
|
|
||||
|
161032
|
Ricca Chemical Company
161032 |
1 L | Natural Poly Bottle |
Each for $50.23
|
|
||||
|
16101
|
Ricca Chemical Company
RI16101 |
4 L | Natural Poly Bottle |
Each for $183.63
|
|
||||
|
16105
|
Ricca Chemical Company
RI16105 |
20 L | Cubitainer |
Each for $431.20
|
|
||||
Specifications
| 97.0,1.02,0.11 | |
| 1X | |
| pH Meter and Electrode | |
| Liquid |
| 100.0,1.06,0.11 | |
| Calibration | |
| 11 | |
| pH Buffer |