missing translation for 'onlineSavingsMsg'
Learn More

Buffer, for Fluoride, Citrate - Acetate, Ricca Chemical

$209.50 - $288.13

Specifications

Color Colorless
Physical Form Liquid
Packaging Natural Poly Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
1478-16
View Documents
Ricca Chemical Company
147816
500 mL
Each for $209.50
Only null left
 
1478-1
View Documents
Ricca Chemical Company
14781
4 L
Each for $288.13
Only null left
 
Description

Description

Specifications

Specifications

Colorless
Liquid
Natural Poly Bottle
Videos
SDS
Documents

Documents