missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Genetrodes™ Electrodes

Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.

Supplier:  BTX™ 450113

 View more versions of this product

Catalog No. BTX508


Only null left
Explore available promotions

Description

Description

  • Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Five models available
  • Models 508 and 510 are straight with gold tips designed for general in vivo applications
  • Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation

Order Info

Genetrode holder with shaft available separately (Cat. No. BTX515).

Specifications

Specifications

In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery
Gold
Electroporation accessory
Genetrodes, 5 mm Straight (GOLD TIP)
Genetrode only
5 mm
1 sec. to 99 msec.
1 Ea.
1 to 10 mm
Non-sterile
Digital User
ECM 830, ECM 2001+, ECM 2001, and Gemini X2
BTX Electroporators
200 V
Straight
0.19 in.
∼1500+ Pulses
10 μsec. to 99 msec. in PBS
0.03 to 0.39 in.
Gold Tip
Banana Plug to Micrograbber
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Intended for Research use only. Not for use in diagnostic or clinical procedures.