missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Caliper Electrodes
Caliper Electrodes for in-vivo electroporation.
Supplier: BTX™ 45 0101
Description
- For in vivo electroporation, specifically the delivery of molecules or drugs into a desired tissue
- Consist of two stainless-steel electrodes that grasp the surface site to be electroporated
- Applications include:
- Electrochemotherapy (ECT): the use of pulsed electric fields to deliver chemotherapeutic drugs into cancerous tumors or
- Transdermal Delivery (TDD): use of a pulsed electric field to deliver drugs or other molecules through the skin to the targeted site
- For use with BTX Gemini X2 electroporation systems.
Chemical Identifiers
| 500 V | |
| Unlimited when properly maintained | |
| 0.039 to 5.11 in. | |
| Non-sterile |
| Square | |
| 10 μsec. to 99 msec. | |
| 1 to 130 mm | |
| Banana Cables |
Specifications
| In-Vivo Drug or Gene Delivery, Transdermal Applications, Intact Plant Applications, Electrochemotherapy | |
| Brass | |
| Caliper Electrode , 1.0 cm X 1.0 cm, Brass | |
| 0.39 in. | |
| Unlimited when properly maintained | |
| 1 Ea. | |
| 1 to 130 mm | |
| 0.39 in. | |
| Non-sterile | |
| Exponential Decay Wave, Square Wave |
| ECM630, ECM 830, Gemini X2, ECM 2001+, and Legacy ECM 2001 Electroporation Systems | |
| 500 V | |
| Square | |
| 1 cm | |
| 10 μsec. to 99 msec. | |
| 0.039 to 5.11 in. | |
| Brass, square | |
| 1 cm | |
| Banana Cables |
Intended for Research use only. Not for use in diagnostic or clinical procedures.