missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
Supplier: BTX™ 450467
Description
- 25-well disposable electroporation plate
- Standard multi-well formats
- Bio-compatible materials
Specifications
| siRNA Transfection, Library Studies and Stem Cell Project | |
| Medical grade polycarbonate, polished aluminium | |
| Electroporation consumables | |
| 25-well Electroporation Plate | |
| 6 Pk. | |
| 2 mm | |
| Sterile | |
| Yes |
| ECM 630HT, ECM830HT, Gemini X2 HT Electroporation System | |
| BTX Electroporation | |
| 25-Well Disposable Electroporation Plate, 2 mm gap, 100 μl, pkg. of 6 | |
| 6 plates plus seals | |
| 0.078 in. | |
| 100 μL (Working) | |
| Exponential Decay Wave, Square Wave |