Learn More
BTX™ HT 25-well Electroporation Plates
HT 25-well Electroporation Plates for use with ECM 630 and ECM 830 Electroporation Systems.
$272.96 - $1525.06
Specifications
Product Line BTX Electroporation
Product Type Electroporation consumables
Electroporation Chamber 25-well Electroporation Plate
Sterility Sterile
High-throughput Compatibility Yes
View More Specs
Products 2
Catalog Number Mfr. No. Size (Metric) Gap Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Size (Metric) Gap Volume (Metric) Quantity Price Quantity  
BTXHTP252
View Documents
BTX™
450466
2 mm 100 μL (Working) 1 Ea.
Each for $272.96
Only null left
BTX450467
View Documents
BTX™
450467
2 mm 100 μL (Working) 6 Pk.
Pack of 6 Each for $1,525.06
Only null left
Description
Description
  • 25-well disposable electroporation plate
  • Standard multi-well formats
  • Bio-compatible materials
Specifications
Specifications
BTX Electroporation
25-well Electroporation Plate
Yes
Electroporation consumables
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'