missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BT-549 Control Lysate (1mg/mL)

Catalog No. PIA37962
Change view
Click to view available options
Quantity:
0.5 mg (1 mg/mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA37962 0.5 mg (1 mg/mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA37962 Supplier Thermo Scientific™ Supplier No. A37962
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ BT-549 Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the TP53 signaling pathway.

  • Validated for the expression of TP53 and phosphorylated TP53 by western blotting and mass spectrometry analysis
  • Optimized - stimulation and lysis conditions optimized for detection of TP53 signaling targets
  • Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
  • Stable for two years at -80°C

Specifications

Detection Method Mass Spectrometry
For Use With (Equipment) Mass Spectrometer
Shipping Condition Dry Ice
Workflow Step Sample Enrichment
Form Liquid
Product Line Thermo Scientific
Quantity 0.5 mg (1 mg/mL)
Type Control Lysate
Final Product Type Protein
Starting Material Cell Lysate
What are the characteristics of BT-549 Control Lysate, and what is its main purpose?

BT-549 Control Lysate is a stimulated BT-549 cell lysate that has been validated for the expression levels and phosphorylation of key targets (TP53 and phosphorylated TP53) in the TP53 signaling pathway. It serves as a positive control for use with SureQuant TP53 panel kits.

For Research Use Only. Not for use in diagnostic procedures.