Learn More
Brucine
CAS: 357-57-3 | C23H26N2O4 | 394.471 g/mol
Supplier: Thermo Scientific Chemicals J6117814
Description
Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBrucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.
Solubility
Soluble in alcohol, chloroform, and benzene, slightly soluble in water, ether, and glycerol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| Brucine | |
| 175°C to 178°C (decomposition) | |
| C23H26N2O4 | |
| 25 g | |
| 63046 | |
| brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
| RRKTZKIUPZVBMF-VLTSECPOSA-N | |
| COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC | |
| 394.471 | |
| 394.47 |
| 357-57-3 | |
| White | |
| MFCD00005942 | |
| UN1570 | |
| 14,1455 | |
| Soluble in alcohol,chloroform,and benzene,slightly soluble in water,ether,and glycerol | |
| −118° (c=2.5 in Chloroform) | |
| (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one | |
| 51413923 | |
| Powder |
Chemical Identifiers
| 357-57-3 | |
| 394.471 | |
| RRKTZKIUPZVBMF-VLTSECPOSA-N | |
| 51413923 | |
| COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC |
| C23H26N2O4 | |
| MFCD00005942 | |
| brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy | |
| (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
H300+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BRUCINE
EINECSNumber : 206-614-7
RTECSNumber : EH8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only