missing translation for 'onlineSavingsMsg'
Learn More

Brucine sulfate hydrate, 98% (dry wt.), water <13%

Catalog No. p-7024746
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1994314 25 g
AAA1994322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1994314 Supplier Thermo Scientific Chemicals Supplier No. A1994314
Only null left

CAS: 652154-10-4 | C46H54N4O12S | 887.01 g/mol

Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine sulfate hydrate is used as a tool for stereospecific chemical syntheses.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 652154-10-4
Molecular Formula C46H54N4O12S
Molecular Weight (g/mol) 887.01
MDL Number MFCD00013472
InChI Key HCMSIGALSOEZRW-MDQLRNFDSA-N
Synonym brucine sulfate salt
PubChem CID 118797651
IUPAC Name 10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;sulfuric acid;hydrate
SMILES OS(O)(=O)=O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C([C@@]35[H])[C@]1(CCN2C4)C1=C6C=C(OC)C(OC)=C1

Specifications

Melting Point 191°C to 196°C (decomposition)
Quantity 25 g
UN Number UN2811
Sensitivity Hygroscopic
Merck Index 14,1455
Solubility Information Soluble in water.
Formula Weight 443.52 (Anhydrous)
Percent Purity 98%
Assay (dry wt.), water <13%
Chemical Name or Material Brucine sulfate hydrate
Hazard Category H300+H330
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 225-432-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only