missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brucine Hydrochloride Monohydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 1544 |
| Formula Weight | 430.93 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Brucine Hydrochloride Monohydrate |
Safety and Handling
| TSCA | Yes |